Learn More
Invitrogen™ TUBA4A Monoclonal Antibody (KT204)
GREENER_CHOICE
Código de producto. 17939102
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
17939102 100 μL 100 microgramos
1 options
Código de producto. 17939102
Marca: Invitrogen™ MA549493
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
This product was used in an Indirect ELISA.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa, are homologous but not identical. The alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TUBA4A
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Western Blot
Clasificación Monoclonal
Clon KT204
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
génica TUBA4A
N.º de referencia del gen P68366
Alias de gen Alpha-tubulin 1; Alpha-tubulin 4; alpha-tubulin isotype M-alpha-4; ALS22; H2-ALPHA; M[a]4; Testis-specific alpha-tubulin; TUBA1; Tuba4; TUBA4A; Tubulin alpha; tubulin alpha 4a; tubulin alpha-1 chain; Tubulin alpha-4 chain; tubulin alpha-4A chain; Tubulin H2-alpha; tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a
Símbolos de los genes TUBA4A
Especie del huésped Mouse
Inmunógeno Recombinant SARS-CoV-2 Nucleocapsid Protein.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7277
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ