Learn More
SPAG5 Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
Descripción
A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.
Especificaciones
Especificaciones
| Antígeno | SPAG5 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3F9 |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | SPAG5 |
| N.º de referencia del gen | Q96R06 |
| Alias de gen | DEEPEST, MAP126, hMAP126 |
| Símbolos de los genes | SPAG5 |
| Mostrar más |