Learn More
Invitrogen™ Kit de ELISA de lisado celular LIX (CXCL5) para ratas
Description
Includes
Placas precubiertas, estándar, anticuerpo detector, conjugado HRP, diluyentes, tampón de lavado, cromógeno, solución de parada y cubiertas de placa.
The Rat LIX (CXCL5) ELISA quantitates Rt LIX in rat cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Rt LIX. Principle of the method The Rat LIX solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Número de acceso | P97885 |
| Intervalo del ensayo | De 8,23 pg/ml a 6000 pg/ml |
| Sensibilidad del ensayo | 15 pg/ml |
| Intervalo de calibración | De 15 pg/ml a 6000 pg/ml |
| ensayo | ELISA sandwich de fase sólida |
| Conjugado | Biotina |
| Tipo de producto | Kit de ELISA |
| Familia de proteínas | Quimiocinas y receptores |
| Tipo de muestra | Lisados celulares |
| Especificidad | El par de anticuerpos suministrado en este kit reconoce LIX/CXCL5 de rata. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.