Learn More
Invitrogen™ ETV1 Polyclonal Antibody
GREENER_CHOICE
Código de producto. 16507161
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16507161 100 μL 100 microlitros
1 options
Código de producto. 16507161
Marca: Invitrogen™ PA589150
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Rabbit Polyclonal Antibody
Immunogen sequence: AFHGLPLKIK KEPHSPCSEI SSACSQEQPF KFSYGEKCLY NVSAYDQKPQ VGMRPSNPPT PSSTPVSPLH HASPNSTHTP KPDRAFPAHL PPSQSIPDSS YPMDHRFRRQ LSEPCNSFPP LPTMPREGRP MYQRQMSEPN IPFPPQGFKQ EYHDPVYEHN TMVGSAASQS FPPPLMIKQE PRDFAYDSEV PSCHSIYMRQ EGFLAHPSRT EGCMFEKGPR Q.
ETV1: ets variant gene 1. Several members of the Ets gene family encode sequence-specific DNA binding proteins that recognize DNA sequences with a centrally located 5'-GGAA-3' element. All of the Ets proteins recognize the same central core sequence but each protein interacts with unique sequences that flank this core. ER81 (also designated ETV1) binds the motif 5'-CGGAA/T-3'. ER81 is highly expressed in brain, testis, lung and heart. ER81 is also moderately expressed in spleen, pancreas, colon and small intestine. During development, ER81 displays a unique expression patterns which suggests that this transcriptional factor might play an important role in organogenesis. ERK-1 activates ER81 transcriptional activity while MAPKAP kinase 2 inhibits ER81.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno ETV1
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.97 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica Etv1
N.º de referencia del gen P41164, P50549
Alias de gen DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein
Símbolos de los genes Etv1
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 80-300 of human ETV1 (NP_0049472).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14009, 2115, 362733
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG