Learn More
Temozolomide, MedChemExpress
Descripción
Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. Temozolomide has antitumor and antiangiogenic effects.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Temozolomide |
| CAS | 85622-93-1 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C6H6N6O2 |
| Cantidad | 200 mg |
| Sinónimo | NSC 362856 CCRG 81045 TMZ |
| Información de solubilidad | DMSO : 20.83 mg/mL (107.29 mM; Need ultrasonic) ∣H2O : 2.86 mg/mL (14.73 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(N2C=N1)N=NN(C)C2=O)N |
| Peso molecular (g/mol) | 194.15 |
| Mostrar más |