Learn More
Invitrogen™ ZIP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA564987
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: TASATFESARN LAIGIGIQNF PEGLAVSLPL RGAGFSTWRA FW Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
ZIP11, also known as Slc39A11, is a member of the broader ZIP family of multi-transmembrane domain metal ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Members of the ZIP family generally transport metal ions from the cell exterior or lumen of intracellular organelles into the cytoplasm, as opposed to the ZnT (SLC30) family of zinc transporters that serve to reduce intracellular zinc availability by promoting zinc efflux from cells or into intracellular vesicles. At least three isoforms of ZIP11 are known to exist.
Especificaciones
| ZIP11 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A11 | |
| 1810074D23Rik; C17orf26; SLC39A11; solute carrier family 39 (metal ion transporter), member 11; solute carrier family 39 member 11; solute carrier family 39, member 11; zinc transporter ZIP11; ZIP11; ZIP-11; Zrt- and Irt-like protein 11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 201266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1S5 | |
| SLC39A11 | |
| Recombinant Human ZIP11. Recombinant protein control fragment (Product #RP-105749). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction