Learn More
anti-IGF-II R/Mannose 6 Phosphate Receptor (Cation independent), Clone: 2G11, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Marca: Novus Biologicals NB300-514
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.Especificaciones
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, CyTOF-ready | |
| P08169 | |
| IGF2R | |
| Purified bovine Mannose 6 Phosphate Receptor (Cation independent) [UniProt# P08169] | |
| 0.1 mL | |
| Primary | |
| Does not react with Chinese hamster ovary cell Mannose 6 Phosphate Receptor. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 2G11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3482 | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |