Learn More
Invitrogen™ Human JPH4 Synthetic Peptide
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Descripción
JPH4 Synthetic Peptide, PEP-0766, from Invitrogen.
Junctional complexes between the plasma membrane (PM) and endoplasmic/sarcoplasmic reticulum (ER/SR) are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. Junctophilins (JPs) are important components of the junctional complexes. JPs are composed of a carboxy-terminal hydrophobic segment spanning the ER/SR membrane and a remaining cytoplasmic domain that shows specific affinity for the PM. Four JPs have been identified as tissue-specific subtypes derived from different genes: JPH1 is expressed in skeletal muscle, JPH2 is detected throughout all muscle cell types, and JPH3 and JPH4 are predominantly expressed in the brain. In the CNS, both JPH3 and JPH4 are expressed throughout neural sites and contribute to the subsurface cistern formation in neurons. Mice lacking both JPH3 and JPH4 subtypes exhibit serious symptoms such as impaired learning and memory and are accompanied by abnormal nervous functions.
Especificaciones
Especificaciones
| Gene ID (Entrez) | 84502 |
| Especie | Human |
| Método de purificación | purified |
| Número de acceso | Q96JJ6 |
| Nombre común | Human JPH4 |
| Concentración | 200 μg/mL |
| Contenido y almacenamiento | -20°C |
| Descripción | Synthetic Peptide |
| Formato | Liquid |
| Formulación | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Mostrar más |