Learn More
Invitrogen™ S100B Monoclonal Antibody (SP127)
GREENER_CHOICE
Código de producto. 13461127
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
500 microlitros
Código de producto. Cantidad unitSize
13461127 500 μL 500 microlitros
1 options
Código de producto. 13461127
Marca: Invitrogen™ MA516396
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Rabbit Monoclonal Antibody
A suggested positive control is colon or melanoma tissue.
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno S100B
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon SP127
Concentración 0.22 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA and 0.1% sodium azide; pH 7.6
génica S100B
N.º de referencia del gen P04271
Alias de gen AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P
Símbolos de los genes S100B
Especie del huésped Rabbit
Inmunógeno Full length recombinant human S100 beta.
Método de purificación Protein A/G
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6285
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG