Learn More
Invitrogen™ O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (9D1.E4)
GREENER_CHOICE
Código de producto. 13247638
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
13247638 100 μg 100 microgramos
1 options
Código de producto. 13247638
Marca: Invitrogen™ MA1039
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
MA1-039 has been successfully used in Western blot and immunoprecipitation applications with human samples. In Western blot, MA1-039 detects proteins with O-GlcNAc post-translational modification, and therefore will detect multiple specific bands.
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno O-linked N-acetylglucosamine (O-GlcNAc)
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon 9D1.E4
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica OGT
N.º de referencia del gen O15294
Alias de gen GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase
Símbolos de los genes OGT
Especie del huésped Mouse
Inmunógeno SANM-conjugated linear peptide corresponding to human O-GluNAc.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8473
Especies diana Chemical
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1