Learn More
Alambre de acero inoxidable, 0,127 mm de diámetro, tipo 304, Thermo Scientific Chemicals
Descripción
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Nombre del producto químico o material | Stainless Steel wire, Type 304 |
| Nombre de nota | 0.127mm (0.005 in.) dia. |
| Peso Mol. medio o Intervalo de pesos mol. | Fe:Cr:Ni; 70:19:11 wt% |
| Cantidad | 50 cm |
| Olor | Odorless |
| Formulario | Alambre |
| Número MDL | MFCD00148499 |
| Información de solubilidad | Insoluble in water. |
Safety and Handling
- Stainless steel wire, type 304
- Peligro
- Carcinogenicidad Categoría 2
- Sensibilización cutánea Categoría 1
- Toxicidad específica para determinados órganos Categoría 1
- H317-Puede provocar una reacción alérgica en la piel.
- H351-Se sospecha que provoca cáncer.
- H372-Provoca daños en los órganos.
- P201-Pedir instrucciones especiales antes del uso.
- P280-Llevar guantes/prendas/gafas/máscara de protección.
- P302+P352-EN CASO DE CONTACTO CON LA PIEL: Lavar con abundante agua/.
- P308+P313-EN CASO DE exposición manifiesta o presunta: Consultar a un médico.
- MIXTURE LIST-Contém: Nickel
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.