Learn More
Invitrogen™ LXR alpha Monoclonal Antibody (K8607)
GREENER_CHOICE
Código de producto. 10328323
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10328323 100 μL cada uno
1 options
Código de producto. 10328323
Marca: Invitrogen™ 418200
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
This antibody specifically recognizes LXR alpha but does not recognize human LXR beta.
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiquitination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LXR alpha
Aplicaciones ChIP Assay, ELISA, Gel Shift, Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon K8607
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación Physiological saline with 0.1% sodium azide
génica NR1H3
N.º de referencia del gen Q13133
Alias de gen AU018371; liver X nuclear receptor alpha variant 1; liver X receptor alpha; LXR; LXR alpha; Lxra; LXR-a; LXRalpha; LXRb; NER; NERI; NR1H2; Nr1h3; nuclear orphan receptor LXR-alpha; nuclear oxysterol receptor LxR-alpha; nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; oxysterols receptor LXR-alpha; RLD1; RLD-1; ubiquitously-expressed nuclear receptor 1; Unr1
Símbolos de los genes NR1H3
Especie del huésped Mouse
Inmunógeno Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant human LXR--alpha (2-97 aa).
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10062
Especies diana Human
Contenido y almacenamiento Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a