missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: APARDELPDW NEVLPPWDRE EDEVYPPGPY HPFPNYIRPR TLQPPSALRR RHYHHALPPS RHYPGREAQA RRAQEEAEAE ERRLQEQEEL ENYIEHV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
VGF was initially identified as a rapidly regulated gene product in nerve growth factor-treated PC12 cells. This protein is synthesized in neurons in the central and peripheral nervous system as well as in the pituitary, adrenal medulla, endocrine cells of the stomach, and pancreatic beta cells. VGF is thought to be involved in organism energy balance and regulation of homeostasis as mice lacking this gene show deficits in these areas. More recent results suggest that VGF is upregulated by brain-derived neurotrophic factor (BDNF) and can stimulate the proliferation of hippocampal progenitor cells and produce antidepressant-like behavioral effects, suggesting that this pathway may be a suitable target for therapeutic treatments.
Especificaciones
Especificaciones
| Antígeno | VGF |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.10 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | VGF |
| N.º de referencia del gen | O15240 |
| Alias de gen | Antimicrobial peptide VGF[554-577]; Gm1052; NERP-1; NERP-2; Neuroendocrine regulatory peptide-1; Neuroendocrine regulatory peptide-2; neuro-endocrine specific protein VGF; Neurosecretory protein VGF; SCG7; SgVII; Vgf; VGF nerve growth factor inducible; VGF(180-194); VGF(24-63); VGF(375-407); VGF8a protein; VGF-derived peptide AQEE-30; VGF-derived peptide HFHH-10; VGF-derived peptide LQEQ-19; VGF-derived peptide TLQP-11; VGF-derived peptide TLQP-21; VGF-derived peptide TLQP-30; VGF-derived peptide TLQP-62 |
| Símbolos de los genes | VGF |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
