missing translation for 'onlineSavingsMsg'
Learn More

VersiCap™ Mat Cap Strips PROMO

Flexible mats of flat, domed cap strips for PCR or ultra clear cap strips for qPCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

Marca:  Thermo Scientific ABgene AB1810100-RMD142

Código de producto. 15679434

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

The Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Highlights

  • Multiple cap strips can be applied at the same time when sealing qPCR plates, thereby reducing setup time
  • Remove the need to cut up adhesive seals
  • Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
  • PCR
  • qPCR/Real-Time PCR

Autoclavable Resistance: Successfully Tested
DMSO (100%) Resistance: Successfully Tested
Ethanol Resistance: Successfully Tested
Gamma Irradiation Resistance: Successfully Tested
Hazardous: No
Isopropanol Resistance: Successfully Tested
Long Term Storage: Successfully Tested
PCR (including waterbath): Successfully Tested
Peelable: Successfully Tested
Pierceable: Not Recommended
qPCR:Ultra Clear caps: Successfully Tested
Domed or Flat caps: Not Recommended
Resealable: Successfully Tested
Sealing Temperature Range: −20°C to +120°C
Shelf Life: 60 Months
Shipping Condition: Ambient
Storage Condition: Ambient
UV Irradiation Resistance: Successfully Tested

  • PCR
  • qPCR/Real-Time PCR

TRUSTED_SUSTAINABILITY
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.