missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veliparib, MedChemExpress
Description
Veliparib (ABT-888) is a potent PARP inhibitor, inhibiting PARP1 and PARP2 with Kis of 5.2 and 2.9 nM, respectively.
Specifications
Specifications
| Nombre del producto químico o material | Veliparib |
| Forma física | Solid |
| CAS | 912444-00-9 |
| Color | Off-White |
| Cantidad | 100 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C13H16N4O |
| Para utilizar con (aplicación) | Cancer-programmed cell death |
| Sinónimo | ABT-888 |
| Información de solubilidad | DMSO : ≥ 29 mg/mL (118.71 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?