Learn More
VDAC1/Porin Rabbit Recombinant Antibody, Proteintech
Código de producto. 30233889 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30233889 20 μL 20 microlitros
30236072 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30233889 Proveedor Proteintech N.º de proveedor 815381RR20UL
Rabbit Recombinant Antibody
The VDAC1/Porin antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human VDAC1/Porin. This antibody recognizes human, mouse, rat antigen. The VDAC1/Porin antibody has been validated for the following applications: WB, IHC, IF-P, FC (Intra), ELISA analysis.
Especificaciones
Antígeno VDAC1/Porin
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Clasificación Recombinant Monoclonal
Clon 2F4
Concentración 500 μg/mL
Conjugado Unconjugated
Símbolos de los genes MGC111064; PORIN; PORIN-31-HL
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Protein A purification
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7416
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 31 kDa
Tipo de producto Recombinant Antibody
Formulario Liquid
Isotype IgG