Learn More
R&D Systems™ VasoTACS In Situ Apoptosis Detection Kit
Código de producto. 18442010
Click to view available options
Cantidad:
30 pruebas
Tamaño de la unidad:
30 tests
Código de producto. 18442010
Marca: R&D Systems™ 482630K
Includes
Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 3mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2
Provides all the reagents for the detection of DNA fragmentation among vascular cells and tissue sections using TUNEL
Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform
Contain TACS BlueLabel, Red Counterstain C, and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in vascular tissues
Study fixed cells, embedded tissues and frozen tissues using light microscopy
VasoTACS
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 30 pruebas
Contenido y almacenamiento The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.