Learn More
R&D Systems™ VasoTACS In Situ Apoptosis Detection Kit
Descripción
Includes
Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 3mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2
Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform
Contain TACS BlueLabel, Red Counterstain C, and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in vascular tissues
Study fixed cells, embedded tissues and frozen tissues using light microscopy
VasoTACS
Especificaciones
Especificaciones
| Cantidad | 30 pruebas |
| Contenido y almacenamiento | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.