Learn More
Epredia™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody
Tienda Epredia ProductosDescripción
Host Species: Mouse
Clone: 5C3.F8
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human VEGF from a glioma
Molecular Weight: 19-22kDa (reduced)
Positive Control: VEGF165 recombinant protein
Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-2 does not neutralize the bioactivity of VEGF.
Recommended for:
- Western Blotting (Reduced only)
- Neutralization of VEGF (Not suitable)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Especificaciones
| Antígeno | Vascular Endothelial Growth Factor (VEGF) Ab-2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 5C3.F8 |
| Concentración | 1mg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Human VEGF from a glioma |
| Cantidad | 200 μL |
| Estado normativo | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.