Descripción
The VAPB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VAPB. This antibody recognizes human, mouse, rat antigen. The VAPB antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | VAPB |
| Aplicaciones | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Clasificación | Polyclonal |
| Concentración | 300 μg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 9217 |
| Especies diana | Human, Rat, Mouse |
| Mostrar más |