Learn More
VAPB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16843134 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16843134 150 μL 150 microlitros
16853134 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16843134 Proveedor Proteintech N.º de proveedor 144771AP150UL
Rabbit Polyclonal Antibody
The VAPB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VAPB. This antibody recognizes human, mouse, rat antigen. The VAPB antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
Antígeno VAPB
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry
Clasificación Polyclonal
Concentración 300 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 9217
Especies diana Human, Rat, Mouse
Isotype IgG