Learn More
VAPA Mouse, anti-Human, Unconjugated, Clone: 1F10F9, Proteintech
Código de producto. 17291506 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
17291506 20 μL 20 microlitros
17281506 150 μL 150 microlitros
2 options
Código de producto. 17291506
Marca: Proteintech 678451IG20UL
Mouse Monoclonal Antibody
VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.
Especificaciones
Antígeno VAPA
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Monoclonal
Clon 1F10F9
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 9218
Especies diana Human, Mouse, Rat