missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vadadustat, MedChemExpress
Description
Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo.
Specifications
Specifications
| Nombre del producto químico o material | Vadadustat |
| CAS | 1000025-07-9 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C14H11ClN2O4 |
| Cantidad | 100 mg |
| Sinónimo | PG-1016548 AKB-6548 |
| Información de solubilidad | DMSO : 100 mg/mL (326.05 mM; Need ultrasonic) |
| SMILES | O=C(O)CNC(C1=NC=C(C2=CC=CC(Cl)=C2)C=C1O)=O |
| Peso molecular (g/mol) | 306.7 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?