missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAChT Polyclonal Antibody

Descripción
Keep as concentrated solution. Predicted reactivity: Human (84%), Mouse (100%), Rat (98%). Positive Control: mouse eye. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Especificaciones
Especificaciones
| Antígeno | VAChT |
| Aplicaciones | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.83 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| génica | SLC18A3 |
| N.º de referencia del gen | O35304, Q62666 |
| Alias de gen | MGC12716; rVAT; SLC18A3; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; solute carrier family 18 (vesicular monoamine), member 3; solute carrier family 18 member 3; solute carrier family 18 member A3; solute carrier family 18, member 3; VAChT; VAT; vesicular acetylcholine transporter |
| Símbolos de los genes | SLC18A3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?