missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaborbactam, MedChemExpress
Descripción
Vaborbactam (RPX7009) is a cyclic boronic acid pharmacophore β-lactamase inhibitor.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Vaborbactam |
| Forma física | Solid |
| CAS | 1360457-46-0 |
| Color | Gray |
| Cantidad | 50 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C12H16BNO5S |
| Para utilizar con (aplicación) | COVID-19-immunoregulation |
| Sinónimo | RPX7009 |
| Información de solubilidad | H2O : 5.26 mg/mL (17.70 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?