Learn More
BIOTIX™ uTIP™ Non-Sterile Low Retention Universal Pipette Tips, Standard
GREENER_CHOICE
Código de producto. p-7146919
Change view
Click to view available options
Volumen (métrico):
250 μL
1000 μL
Tamaño de la unidad:
paquete de 960
Código de producto. Volumen (métrico) unitSize
30216548 250 μL paquete de 960
30216561 1000 μL paquete de 960
2 options
Código de producto. 30216548
Marca: BIOTIX™ 63300058
Designed with FlexFit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Non-Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.
  • Compatible with the widest range of industry leading pipettes - Fits all standard pipettes
  • Accurate and precise channel-channel draw on multichannel pipettes
  • Secure seal on single and multichannel models
  • FlexFit™ seals on your pipette with less force, reducing your ejection force
  • FlexFit™ technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin™ maximizes your sample recovery by creating a low binding surface for DNA, proteins, and nanoparticles - resulting in tighter CV values
  • Blade™ lowers chance of hanging droplet formation by optimizing surface area at distal end of tip - ensures a clean delivery of your sample, increasing accuracy and precision
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
  • Better reprodicibility
  • Certifications
    ISO 9001
    Especificaciones
    Descripción 250 μL, Racked
    Volumen (métrico) 250 μL
    filtradas por Sin filtro
    Esterilidad No estéril
    Color Clear
    Color de gradilla Verde
    Esterilizables en autoclave Autoclavable
    Modelo Universal
    Longitud (métrico) 59.54 mm
    Certificaciones/conformidad ISO 9001
    Declaraciones de limpieza Reusable as a 96 well reservoir
    Para utilizar con (aplicación) Low binding for Protein, DNA, and nanoparticles
    Formato Hinged Rack
    Estilo de punta Ergonomic
    Envase En rack
    Cantidad 10 racks/pk
    Tipo de producto Pipette Tips
    Desechables Single-use
    Para utilizar con (equipo) Universal Fit
    Graduado Sí
    Línea de productos uTip
    Material Polipropileno
    Forma Conical
    Graduaciones 10 μL, 50 μL, 100 μL, 200 μL