Learn More
Uteroglobin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16856903 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16856903 150 μL 150 microlitros
16866903 20 μL 20 microlitros
2 options
Código de producto. 16856903
Marca: Proteintech 104901AP150UL
Rabbit Polyclonal Antibody
Uteroglobin encodes a member of the secretoglobin family of small secreted proteins. The encoded protein has been implicated in numerous functions including anti-inflammation, inhibition of phospholipase A2 and the sequestering of hydrophobic ligands. Defects in this gene are associated with a susceptibility to asthma.
Especificaciones
Antígeno Uteroglobin
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SCGB1A1
N.º de referencia del gen P11684, Q06318
Alias de gen CC10, CC16, CCPBP, CCSP, SCGB1A1, UGB, UP 1, UP1, Urinary protein 1, Urine protein 1, Uteroglobin
Símbolos de los genes SCGB1A1
Especie del huésped Rabbit
Inmunógeno Uteroglobin Fusion Protein Ag0758
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 22287, 7356
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG