missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Uteroglobin Monoclonal Antibody (3A8B8)

Descripción
MA5-17170 targets SCGB1A1 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17170 immunogen is purified recombinant fragment of human SCGB1A1 (amino acids: 26-91) expressed in E. Coli. MA5-17170 detects SCGB1A1 which has a predicted molecular weight of approximately 10kDa.
Uteroglobin, which is a member of the Secretoglobin superfamily and is also known as Clara cell phospholipid-binding protein, is a multifunctional protein that can exert anti-inflammatory and anti-tumorigenic effects by binding small hydrophobic molecules such as phospholipids and prostaglandins. The small, non-glycosylated protein named for its high levels of expression in pre-implantation embryos, where it exhibits growth stimulatory effects, is produced and secreted by the non-ciliated, non-mucous Clara cells predominant in the epithelial surfaces of pulmonary airways, as well as other non-ciliated epithelia. Members of the Secretoglobin superfamily demonstrate a high level of structural conservation and are characterized as small, secretory homo- or heterodimers. In addition to sequestering pro-inflammatory mediators and carcinogens, Uteroglobin has been implicated in the inhibition of cell migration and invasion, platelet aggregation, and T cell differentiation.
Especificaciones
Especificaciones
| Antígeno | Uteroglobin |
| Aplicaciones | ELISA, Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | 3A8B8 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% sodium azide |
| génica | SCGB1A1 |
| N.º de referencia del gen | P11684 |
| Alias de gen | Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?