missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USP8 Polyclonal Antibody

Descripción
USP8 Polyclonal Antibody for IHC (P), Western Blot
USP8 is a ubiquitin specific protease that plays an important regulatory role at the level of protein turnover by preventing degradation. USP8 is involved in cell proliferation, and probably regulates the stability of STAM2 and RASGRF1. USP8 may regulate T-cell energy mediated by RNF128 via the formation of a complex containing RNF128 and STAM2. As revealed by structure/function studies, USP8 forms a ternary complex with RNF128 and OTUB1, and interacts with the SH3 domain of STAM2 and RASGRF1. Expression of USP8 is induced Upon growth stimulation in starved human fibroblasts, and expression decreases in response to growth arrest induced by cell-cell contact.
Especificaciones
Especificaciones
| Antígeno | USP8 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 2.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| génica | USP8 |
| N.º de referencia del gen | P40818, Q80U87 |
| Alias de gen | AI574262; AW557536; Deubiquitinating enzyme 8; hUBPy; HumORF8; Kiaa0055; mKIAA0055; mUBPy; putative deubiquitinating enzyme; SPG59; Ubiquitin carboxyl-terminal hydrolase 8; ubiquitin isopeptidase Y; ubiquitin specific peptidase 8; ubiquitin specific protease 8; ubiquitin thioesterase 8; ubiquitin thiolesterase 8; ubiquitin-specific-processing protease 8; Ubpy; USP8 |
| Símbolos de los genes | USP8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?