Descripción
DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
Especificaciones
Especificaciones
| Antígeno | USP33 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | USP33 |
| N.º de referencia del gen | Q8R5K2, Q8TEY7 |
| Alias de gen | Deubiquitinating enzyme 33, hVDU1, KIAA1097, Ubiquitin thiolesterase 33, USP33, VDU1 |
| Símbolos de los genes | Usp33 |
| Mostrar más |