Learn More
USP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16892954 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16892954 20 μL 20 microlitros
16882954 150 μL 150 microlitros
2 options
Código de producto. 16892954
Marca: Proteintech 143541AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the ubiquitin specific protease (USP) family of deubiquitinating enzymes. USP enzymes play critical roles in ubiquitin-dependent processes through polyubiquitin chain disassembly and hydrolysis of ubiquitin-substrate bonds. The encoded protein associates with the COP9 signalosome, and also plays a role in transforming growth factor beta signaling through deubiquitination of receptor-activated SMAD transcription factors. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 2.
Especificaciones
Antígeno USP15
Aplicaciones Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.44 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica USP15
N.º de referencia del gen Q8R5H1, Q9R085, Q9Y4E8
Alias de gen Deubiquitinating enzyme 15, KIAA0529, Ubiquitin thiolesterase 15, Unph 2, UNPH4, USP15
Símbolos de los genes Usp15
Especie del huésped Rabbit
Inmunógeno USP15 Fusion Protein Ag5703
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14479, 171329, 9958
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG