Learn More
Invitrogen™ UQCRFS1 Recombinant Rabbit Monoclonal Antibody (24GB2460)
Código de producto. 30275032
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30275032 20 μL 20 microlitros
30275064 100 μL 100 microlitros
2 options
Código de producto. 30275032
Marca: Invitrogen™ MA557831
Rabbit Recombinant Monoclonal Antibody
UQCRFS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno UQCRFS1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB2460
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica Uqcrfs1
N.º de referencia del gen P20788, P47985, Q9CR68
Alias de gen 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1
Símbolos de los genes Uqcrfs1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human UQCRFS1 (aa 220-274).
Método de purificación Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 291103, 66694, 7386
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG