Learn More
UQCRC2 Mouse anti-Human, Unconjugated, Clone: 1A5G11, Proteintech
Código de producto. 16880236 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16880236 20 μL 20 microlitros
16870236 150 μL 150 microlitros
2 options
Código de producto. 16880236
Marca: Proteintech 675471IG20UL
Mouse Monoclonal Antibody
The UQCRC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UQCRC2. This antibody recognizes human, mouse, rat, pig antigen. The UQCRC2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antígeno UQCRC2
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Monoclonal
Clon 1A5G11
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 7385
Especies diana Human, Mouse, Rat, Pig
Isotype IgG1