Learn More
UPF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28384-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The UPF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human antigen. The UPF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| UPF1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 5976 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human |