missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNKL Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA569227
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This target displays homology in the following species: Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
Especificaciones
| UNKL | |
| Polyclonal | |
| Unconjugated | |
| UNKL | |
| 1300004G08Rik; C16orf28; Grailexp predicted; putative E3 ubiquitin-protein ligase UNKL; RGD1565247; RING finger protein unkempt-like; RING finger protein unkempt-like; putative E3 ubiquitin-protein ligase UNKL; si:ch211-66h3.3; unk like zinc finger; unkempt family like zinc finger; unkempt family zinc finger-like; unkempt homolog (Drosophila)-like; unkempt homolog-like; unkempt-like; unkempt-like (Drosophila); Unkl; ZC3H5L; ZC3HDC5L; Zinc finger CCCH domain-containing protein 5-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 74154 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5FWH2 | |
| UNKL | |
| synthetic peptide directed towards the middle region of mouse UNKL. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido