Learn More
Corning™ Universal Fit Hinged Rack Pipet Tips, Sterile
Tienda Corning Productos
Click to view available options
Volumen (métrico):
De 100 a 1000 μL
Color:
Azul
Tamaño de la unidad:
paquete de 1000
Descripción
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Especificaciones
Especificaciones
| Volumen (métrico) | De 100 a 1000 μL |
| Esterilidad | Estéril |
| Color | Azul |
| Para utilizar con (aplicación) | Hinged closure provides easy stacking and steam penetration during autoclaving |
| Formato | Hinged Rack |
| Estilo de punta | Round |
| Envase | Hinged rack |
| Graduado | Sí |