missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The antibody is specific to UNC5A. It has no cross reaction to UNC5B, UNC5C and UNC5D.
The UNC5H family of proteins act as transmembrane receptors for netrin-1 and play a crucial role in axon guidance and migration of neural cells. Additionally, when cleaved by a caspase to produce an intracellular fragment containing a death domain,UNC5H receptors induce apoptosis. This activity is blocked by the binding of netrin-1. In the absence of netrin-1, UNC5H receptors act as tumor suppressors by inhibiting anchorage-independent growth and invasion, but mutation of these receptors provides a potential mechanism for tumorigenicity. The expression of UNC5H receptors is downregulated in multiple carcinomas, including colorectal, breast, ovary, uterus, stomach, lung and kidney cancers. UNC5H1, also known as UNC5HA (unc-5 homolog A), is a member of the UNC5H family of proteins and is localized to the cell membrane. Three isoforms of UNC5H1 exist due to alternative splicing events.
Especificaciones
Especificaciones
| Antígeno | UNC5A |
| Aplicaciones | Western Blot, Flow Cytometry |
| Clasificación | Polyclonal |
| Concentración | 0.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | UNC5A |
| N.º de referencia del gen | O08721, Q6ZN44, Q8K1S4 |
| Alias de gen | KIAA1976, Netrin receptor UNC5A, Protein unc 5 homolog 1, Protein unc 5 homolog A, unc 5 homolog A (C. elegans), UNC5A, UNC5H1 |
| Símbolos de los genes | Unc5a |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?