missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC2025 hydrochloride, MedChemExpress
Descripción
UNC2025 hydrochloride is a potent, ATP-competitive, and highly orally active Mer/Flt3 inhibitor with IC50 values of 0.74 nM and 0.8 nM, respectively. UNC2025 hydrochloride is >45-fold selectivity for MERTK relative to Axl (IC50= 122 nM
Specifications
Specifications
| Nombre del producto químico o material | UNC2025 hydrochloride |
| CAS | 2070015-17-5 |
| Color | Light Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C28H41ClN6O |
| Cantidad | 50 mg |
| Información de solubilidad | H2O : 55 mg/mL (107.19 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (19.49 mM; Need ultrasonic) |
| SMILES | CN1CCN(CC2=CC=C(C3=CN([C@@H]4CC[C@@H](O)CC4)C5=NC(NCCCC)=NC=C53)C=C2)CC1.[H]Cl |
| Peso molecular (g/mol) | 513.12 |
| Formula Weight (peso de la fórmula) | 513.12 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?