Learn More
UNC13D/Munc13-4 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16816674 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16816674 20 μL 20 microlitros
16806674 150 μL 150 microlitros
2 options
Código de producto. 16816674
Marca: Proteintech 169051AP20UL
Rabbit Polyclonal Antibody
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
Especificaciones
Antígeno UNC13D/Munc13-4
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.23 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica UNC13D
N.º de referencia del gen Q70J99
Alias de gen FHL3, HLH3, HPLH3, Munc13 4, Munc13-4, Protein unc 13 homolog D, unc 13 homolog D (C. elegans), UNC13D
Símbolos de los genes UNC13D
Especie del huésped Rabbit
Inmunógeno UNC13D/Munc13-4 Fusion Protein Ag10519
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 201294
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG