Learn More
Umifenovir hydrochloride, MedChemExpress
Código de producto. 16426596
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
16426596 100 mg 100 miligramos
16486248 10 mg 10 miligramos
16462377 50 mg 50 miligramos
3 options
Código de producto. 16426596
Marca: Medchem Express HY14904A
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
MedChemExpress Umifenovir hydrochloride is a potent, orally active broad-spectrum antiviral with activity against a number of enveloped and non-enveloped viruses. Umifenovir hydrochloride is used as an anti-influenza virus agent. Umifenovir hydrochloride could effectively inhibit the fusion of virus with host cells. Umifenovir hydrochloride is an efficient inhibitor of SARS-CoV-2 in vitro. Anti-inflammatory activity.
Umifenovir hydrochloride is a potent, orally active broad-spectrum antiviral with activity against a number of enveloped and non-enveloped viruses. Umifenovir hydrochloride is used as an anti-influenza virus agent. Umifenovir hydrochloride could effectively inhibit the fusion of virus with host cells. Umifenovir hydrochloride is an efficient inhibitor of SARS-CoV-2 in vitro. Anti-inflammatory activity.
Especificaciones
Nombre del producto químico o material Umifenovir hydrochloride
Forma física Solid
CAS 131707-23-8
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C22H26BrClN2O3S
Para utilizar con (aplicación) COVID-19-anti-virus
Información de solubilidad DMSO : 25 mg/mL (48.65 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=C(CSC2=CC=CC=C2)N(C)C3=C1C(CN(C)C)=C(O)C(Br)=C3)OCC.[H]Cl
Peso molecular (g/mol) 513.88
Formula Weight (peso de la fórmula) 513.88
Porcentaje de pureza 99.68%