missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UGGT1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Código de producto. 17960173
Change view
Click to view available options
Cantidad:
20 μL
200 μL
Tamaño de la unidad:
20 microlitros
200 microlitros
Código de producto. Cantidad unitSize
17960173 200 μL 200 microlitros
17990113 20 μL 20 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17960173

Marca: Invitrogen™ PA5145405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UDP-glucose:glycoprotein glucosyltransferase (UGGT) is a 170 kDa lumenal endoplasmic reticulum (ER) resident protein that catalyzes monoglucosylation of high mannose oligosaccharides. UGGT catalyzes the transfer of glucose from UDP-glucose to Man7-9GlcNAc2-moiety of unfolded glycoproteins in a Ca2+ dependent reaction. UGGT binds unfolded glycoprotein via two elements on its substrate: a N-acetylglucosamine unit on the oligosaccharide and hydrophobic amino acids that are exposed in the denatured conformation. It is postulated that UGGT serves as a sensor that is able to distinguish between folded and denatured protein in the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno UGGT1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
génica Uggt1
N.º de referencia del gen Q9NYU2
Alias de gen 0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr
Símbolos de los genes Uggt1
Especie del huésped Rabbit
Inmunógeno Recombinant Human UGGT1 protein, Leu1221-Leu1536 (Accession #Q9NYU2).
Método de purificación Antigen affinity chromatography, Protein A
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100155117, 56886
Especies diana Human, Pig
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.