Learn More
UGGT1 Polyclonal Antibody
UGGT1 Polyclonal Antibody
Marca: Invitrogen PA5103025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
UGGT1 Polyclonal Antibody for Western Blot
Especificaciones
| UGGT1 | |
| Polyclonal | |
| Unconjugated | |
| Uggt1 | |
| 0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56886 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NYU2 | |
| Uggt1 | |
| A synthesized peptide derived from human UGGT1(Accession Q9NYU2), corresponding to amino acid residues E160-F210. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |