Learn More
UFM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15883-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The UFM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UFM1. This antibody recognizes human, mouse antigen. The UFM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| UFM1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 51569 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |