missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP2 Mouse anti-Human, Clone: 2B4B1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66700-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody detect a band around 28-33 kDa corresponding to UCP2 protein in mouse liver, skeletal muscle and pancreas. Some upper and lower bands also were observed due to the dimer formation and degradation of the UCP2 protein. This antibody was raised
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.Especificaciones
| UCP2 | |
| Monoclonal | |
| 0.83 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P55851 | |
| UCP2 | |
| UCP2 Fusion Protein Ag19361 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| Western Blot | |
| 2B4B1 | |
| Unconjugated | |
| UCP2 | |
| BMIQ4, SLC25A8, UCP 2, UCP2, UCPH | |
| Mouse | |
| Protein G | |
| RUO | |
| 7351 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido