missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCN2 Monoclonal Antibody (C2), Invitrogen™

Descripción
UCN2 (Urocortin 2) is a Protein Coding gene. Diseases associated with UCN2 include Body Mass Index Quantitative Trait Locus 11. Among its related pathways are Signaling by GPCR and Presynaptic function of Kainate receptors. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and G protein-coupled receptor binding. An important paralog of this gene is UCN3. This gene is a member of the sauvagine/corticotropin-releasing factor/urotensin I family. It is structurally related to the corticotropin-releasing factor (CRF) gene and the encoded product is an endogenous ligand for CRF type 2 receptors. In the brain it may be responsible for the effects of stress on appetite. In spite of the gene family name similarity, the product of this gene has no sequence similarity to urotensin II. [provided by RefSeq, Jul 2008].
Especificaciones
Especificaciones
| Antígeno | UCN2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | C2 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | UCN2 |
| N.º de referencia del gen | Q96RP3 |
| Alias de gen | prepro-urocortin 2; SRP; Stresscopin-related peptide; ucn II; UCN2; Ucn-2; Ucn3; UCNI; ucn-II; UR; urocortin 2; urocortin 3; urocortin II; Urocortin-2; urocortin-related peptide; URP |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?