missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ubiquitin C Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17702264
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17702264 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17702264 Proveedor Invitrogen™ N.º de proveedor PA5120862

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive test controls include: 293T, BT-474, HepG2, U-251MG, A-549, Mouse testis, Mouse liver, Rat testis, Rat brain. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MQIFVKTLTG KTITLEVEPS DTIENVKAKI QDKEGIPPDQ QRLIFAGKQL EDGRTLSDYN IQKESTLHLV LRLRGGMQIF VKTLTGKTIT LEVEPSDTIE.

This gene encodes ubiquitin, one of the most conserved proteins known. Ubiquitin is required for ATP-dependent, nonlysosomal intracellular protein degradation of abnormal proteins and normal proteins with a rapid turnover. Ubiquitin is covalently bound to proteins to be degraded, and presumably labels these proteins for degradation. Ubiquitin also binds to histone H2A in actively transcribed regions but does not cause histone H2A degradation, suggesting that ubiquitin is also involved in regulation of gene expression. This gene consists of three direct repeats of the ubiquitin coding sequence with no spacer sequence. Consequently, the protein is expressed as a polyubiquitin precursor with a final amino acid after the last repeat. Aberrant form of this protein has been noticed in patients with Alzheimer's and Down syndrome.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Ubiquitin C
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1.16 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica UBC
N.º de referencia del gen P0CG48, P0CG50, Q63429
Alias de gen 2700054O04Rik; AI194771; HMG20; polyubiquitin C; polyubiquitin-C; Rps27a; TI-225; Uba52; Ubb; Ubc; Ubiquitin; ubiquitin C; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2
Símbolos de los genes UBC
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBC (NP_066289.3).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 22190, 50522, 7316
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.