missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The UBC9-Specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human UBC9-Specific. This antibody recognizes human, mouse, rat antigen. The UBC9-Specific antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | UBC9-Specific |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Monoclonal |
| Clon | 1G8A2 |
| Concentración | 1900 μg/mL |
| Conjugado | Unconjugated |
| Símbolos de los genes | C358B7.1; P18; UBC9 |
| Especie del huésped | Mouse |
| Inmunógeno | Peptide |
| Método de purificación | Protein G purification |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?