missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ TYVEK™ 500 HP Model TY178
Descripción
- Workers operating with full-body harness in environments where hazardous particle or light liquid aerosol hazards are present, previously had to compromise between efficient personal protection and protecting their full-body harness against external contamination.
- The new Tyvek™ 500 HP TY178S HP garment offers a unique solution - protection of both the wearer and the full-body harness - by providing a barrier between the user and the particle or light liquid aerosol hazards in the environment. Designed to be worn over a full-body harness, the innovative, patent-pending garment design allows workers to access and utilize multiple D-Rings on their full-body harness while providing an ideal balance of protection, durability, and comfort.
- This new TY178S HP model has four attached sleeves for use with front, back and side D-Rings of a full-body harness. The off-centered zipper entry allows accommodating the front D-Ring sleeve for front D-Ring connection. Compatible with variety of full-body harness models, this garment may be used for a wide range of applications including painting, cleaning, maintenance, and more.
- In addition to its innovative design, this garment also offers an ideal balance of protection and durability thanks to the unique Tyvek™ fabric. It is also extremely lightweight offering high level of comfort and mobility. The garment is low linting (suitable also for paint applications). Silicone non-added.
Especificaciones
Especificaciones
| Tipo de producto | Collared Coveralls |
| Tamaño | Medium |
| Sexo | Unisex |
| Material | HDPE |
| Color | White |
| Desechables | Single-use |
| Estilo de cintura | Elastic |
| Estilo de tobillo | Elastic |
| Estilo de manguito | Elastic |
| Estilo de cuello | Hooded |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?