missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYK2(phospho Y1054) & TYK2 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DP0182.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein phosphorylation antibody pair set comes with two antibodies, one against the TYK2 protein, and the other against the specific Y1054 phosphorylated site of TYK2 for use in in situ Proximity Ligation Assay.Especificaciones
| TYK2 | |
| Unconjugated | |
| JTK1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-TYK2 Y1054 rabbit polyclonal antibody (20 ul) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. TYK2 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| tyrosine kinase 2 | |
| TYK2 | |
| RUO | |
| 7297 | |
| Human | |
| Ab Pair |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido