Learn More
Saint Gobain Life Sciences™ Tygon™ A-60-G
Click to view available options
Diámetro exterior (imperial):
0.189 in., 4,8 mm
0.378 in., 9,6 mm
Tamaño de la unidad:
15 metros
Descripción
- High-performance tubing is resistant to acids and alkali
- Can withstand extreme temperatures from -75°F to +275°F without cracking or deteriorating, even after years of exposure
- Heat sealable and can be joined without fittings for easy installation
- Available in standard vacuum sizes that will withstand a full vacuum (29.9 mm Hg) at 73°F
- Ideal resistance to weathering and abrasion
- High flexural fatigue resistance
- Low gas permeability versus rubber tubing
- Ozone (300 pphm) and UV light resistant
- Applications include: soap and disinfectant dispensing, cleaning chemical transfer, caustic chemical dispensing, plating and etching chemicals, glass and window wash systems, vacuum pumps
Especificaciones
Especificaciones
| Esterilizables en autoclave | Not Autoclavable |
| Diámetro interior (imperial) | 0,189 in |
| Diámetro interior (métrico) | 4.8 mm |
| Material | Termoplástico |
| Diámetro exterior (imperial) | 0.378 in., 9,6 mm |
| Longitud global (imperial) | 49.2 in. |
| Longitud global (métrico) | 15 m |
| Para utilizar con | High-performance Chemical Dispensing |
| Color | Negro |
| Dureza | 61 (Shore A) |
| Mostrar más |