missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXLNG Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA551375
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KALLQMAEEK TVRDKEYKAL QIKLERLEKL CRALQTERNE LNEKVEVLKE QVSIKAAIKA ANRDLATPVM QPCTALDSHK ELNTSSKRAL GAHLEAEPKS QRSAVQKPPS TGSAP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.
Especificaciones
| TXLNG | |
| Polyclonal | |
| Unconjugated | |
| TXLNG | |
| 4932441K18Rik; CXHXORF15; CXorf15; ELRG; environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; factor inhibiting ATF4-mediated transcription; FIAT; gamma-taxilin; lipopolysaccharide specific response-5 protein; lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; Lrg; Lrp; LSR5; taxilin gamma; TXLNG; TXLNGX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUQ3 | |
| TXLNG | |
| Recombinant protein corresponding to Human TXLNG. Recombinant protein control fragment (Product #RP-91990). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido